missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2G4F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563344
This item is not returnable.
View return policy
Description
Immunogen sequence: LVNRTFRTHL APGVERQTAE EKAFGDFVIN RPDTPYGMMN FTYEPQDFYR LVALSRYNVL NNVETLKCAL QLALDRHQAR ERA Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 42%.
Calcium-dependent phospholipase A2 that selectively hydrolyzes glycerophospholipids in the sn-2 position. Has higher enzyme activity for phosphatidylethanolamine than phosphatidylcholine.
Specifications
| PLA2G4F | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4F | |
| 3.1.1.4; 4732472I07Rik; cPLA2-zeta; cytosolic phospholipase A2 zeta; Gm647; Phospholipase A2 group IVF; phospholipase A2, group IVF; Pla2g4f; PLA2G4FZ; Pla2zeta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 255189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68DD2 | |
| PLA2G4F | |
| Recombinant protein corresponding to Human PLA2G4F. Recombinant protein control fragment (Product #RP-103266). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction