missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19855
This item is not returnable.
View return policy
Description
PLA2G4F Polyclonal specifically detects PLA2G4F in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PLA2G4F | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| PLA2G4F phospholipase A2, group IVF, PLA2G4FZ | |
| Rabbit | |
| 95 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (81%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.8mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q68DD2 | |
| PLA2G4F | |
| Recombinant protein encompassing a sequence within the center region of human PLA2G4F. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 255189 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction