missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13345-1-AP-20UL
This item is not returnable.
View return policy
Description
The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].Specifications
| PLA2G4D | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4D | |
| Cpla2 delta, cPLA2delta, Phospholipase A2 group IVD, PLA2G4D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283748, 78390 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q50L43, Q86XP0 | |
| PLA2G4D | |
| PLA2G4D Fusion Protein Ag4178 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction