missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4D Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00283748-B01P
This item is not returnable.
View return policy
Description
PLA2G4D Polyclonal antibody specifically detects PLA2G4D in Human samples. It is validated for Western Blot
Specifications
| PLA2G4D | |
| Polyclonal | |
| PBS (pH 7.4) | |
| cPLA2delta, cPLA2-delta, cytosolic phospholipase A2 delta, EC 3.1.1.4, phospholipase A2 delta, cytosolic, Phospholipase A2 group IVD, phospholipase A2, group IVD (cytosolic) | |
| PLA2G4D (AAH34571.1, 1 a.a. - 216 a.a.) full-length human protein. MGRLMRRIPEPRICFLEAIWSNIFSLNLLDAWYDLTSSGESWKQHIKDKTRSLEKEPLTTSGTSSRLEASWLQPGTALAQAFKGFLTGRPLHQRSPNFLQGLQLHQDYCSHKDFSTWADYQLDSMPSQLTPKEPRLCLVDAAYFINTSSPSMFRPGRRLDLILSFDYSLSAPFEVPWSPQGNPSAQPGQAPEASSRATEPLPHTARVPKGRRGVRP | |
| 0.05 mg | |
| Endocrinology, Signal Transduction | |
| 283748 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| AAH34571.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction