missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PLA2G4D Polyclonal antibody specifically detects PLA2G4D in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | PLA2G4D |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | cPLA2delta, cPLA2-delta, cytosolic phospholipase A2 delta, EC 3.1.1.4, phospholipase A2 delta, cytosolic, Phospholipase A2 group IVD, phospholipase A2, group IVD (cytosolic) |
| Host Species | Rabbit |
| Immunogen | phospholipase A2, group IVD(cytosolic). (Uniprot# Q86XP0) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?