missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2G4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598062
This item is not returnable.
View return policy
Description
PLA2G4B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
PLA2G4B transcribes naturally-occurring mRNAs that are co-transcribed products of the neighboring JMJD7 and PLA2G4B genes. Incompletely processed read-through transcripts from these two loci are abundantly expressed in most tissues, but the function of the predicted protein product has not yet been determined.
Specifications
| PLA2G4B | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4B | |
| A030011C02Rik; cPLA2-beta; cytosolic phospholipase A2 beta; Gm1013; HsT16992; JMJD7-PLA2G4B; JMJD7-PLA2G4B protein; JMJD7-PLA2G4B readthrough; jumonji domain containing 7-phospholipase A2, group IVB (cytosolic) read-through; phospholipase A2 group IVB; phospholipase A2, group IVB (cytosolic); Pla2g4b | |
| Rabbit | |
| Protein G | |
| RUO | |
| 100137049 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.655 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0C869 | |
| PLA2G4B | |
| Recombinant Human Cytosolic phospholipase A2 beta protein (514-781AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction