missing translation for 'onlineSavingsMsg'
Learn More

PLA2G3 Antibody, Novus Biologicals™

Product Code. 18380919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380919 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380919 Supplier Novus Biologicals Supplier No. H00050487D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLA2G3 Polyclonal antibody specifically detects PLA2G3 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH25316.1
Gene Alias EC 3.1.1.4, GIII sPLA2, GIII-SPLA2, group 3 secretory phospholipase A2, group III secreted phospholipase A2, Group III secretory phospholipase A2, Phosphatidylcholine 2-acylhydrolase 3, phosphatidylcholine 2-acylhydrolase GIII, phospholipase A2, group III, sPLA2-III, SPLA2III
Host Species Rabbit
Immunogen PLA2G3 (AAH25316.1, 1 a.a. - 509 a.a.) full-length human protein. MGVQAGLFGMLGFLGVALGGSPALRWYRTSCHLTKAVPGNPLGYLSFLAKDAQGLALIHARWDAHRRLQACSWEDEPELTAAYGALCAHETAWGSFIHTPGPELQRALATLQSQWEACRALEESPAGARKKRAAGQSGVPGGGHQREKRGWTMPGTLWCGVGDSAGNSSELGVFQGPDLCCREHDRCPQNISPLQYNYGIRNYRFHTISHCDCDTRFQQCLQNQHDSISDIVGVAFFNVLEIPCFVLEEQEACVAWYWWGGCRMYGTVPLARLQPRTFYNASWSSRATSPTPSSRSPAPPKPRQKQHLRKGPPHQKGSKRPSKANTTALQDPMVSPRLDVAPTGLQGPQGGLKPQGARWVCRSFRRHLDQCEHQIGPREIEFQLLNSAQEPLFHCNCTRRLARFLRLHSPPEVTNMLWELLGTTCFKLAPPLDCVEGKNCSRDPRAIRVSARHLRRLQQRRHQLQDKGTDERQPWPSEPLRGPMSFYNQCLQLTQAARRPDRQQKSWSQ
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 50487
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.