missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2G2D Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 16528401
Cambia vista
Click to view available options
Quantity:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
16528401 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16528401 Fornitore Invitrogen™ N. del fornitore PA593147

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

Immunogen sequence: GILNLNKMVK QVTGKMPILS YWPYGCHCGL GGRGQPKDAT DWCCQTHDCC YDHLKTQGCS IYKDYYRYNF SQGNIHCSDK GSWCEQQLCA CDKEVAFCLK RNLDTYQKRL RFYWRPHCRG QTPGC; Positive Samples: HL-60, THP-1.

PA2 catalyzes the calcium-dependent hydrolysis of the 2- acyl groups in 3-sn-phosphoglycerides. L-alpha-1-palmitoyl-2- linoleoyl phosphatidylethanolamine is more efficiently hydrolyzed than the other phospholipids examined.
TRUSTED_SUSTAINABILITY

Specifica

Antigen PLA2G2D
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Pla2g2d
Gene Accession No. Q9UNK4, Q9WVF6
Gene Alias GIID sPLA2; Group IID secretory phospholipase A2; mGIID; phosphatidylcholine 2-acylhydrolase 2D; phospholipase A2 group IID; phospholipase A2, group IID; Pla2a2; PLA2G2D; PLA2IID; secretory phospholipase A2s; secretory-type PLA, stroma-associated homolog; sPLA2-IID; sPLA2S; SPLASH
Gene Symbols Pla2g2d
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18782, 26279, 298579
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.