missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2G2A Recombinant Rabbit Monoclonal Antibody (23GB2190), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552483
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the phospholipase A2 family (PLA2). PLA2s constitute a diverse family of enzymes with respect to sequence, function, localization, and divalent cation requirements. This gene product belongs to group II, which contains secreted form of PLA2, an extracellular enzyme that has a low molecular mass and requires calcium ions for catalysis. It catalyzes the hydrolysis of the sn-2 fatty acid acyl ester bond of phosphoglycerides, releasing free fatty acids and lysophospholipids, and thought to participate in the regulation of the phospholipid metabolism in biomembranes. Several alternatively spliced transcript variants with different 5' UTRs have been found for this gene.
Specifications
| PLA2G2A | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14423, P14555, P31482 | |
| PLA2G2A | |
| A synthesized peptide derived from human PLA2G2A (105-144AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2190 | |
| Unconjugated | |
| PLA2G2A | |
| EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1; non-pancreatic secreted type II phospholipase A2; non-pancreatic secretory phospholipase A2; NPS-PLA2; phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2 group IIA; phospholipase A2, group IIA (platelets, synovial fluid); phospholipase A2, membrane associated; Pla2; PLA2B; PLA2BPLA2; PLA2G2A; PLA2L; PLA2LPLA2S; PLA2S; PLAS1; platelet phospholipase A2; RASF-A; secretory group II phospholipase A2; sPLA2; sPla2-IIA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18780, 29692, 5320 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction