missing translation for 'onlineSavingsMsg'
Learn More

PLA2G2A Rabbit anti-Human, Mouse, Rat, Clone: 3X9F5, Novus Biologicals™

Product Code. 18394785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394785 100 μg 100µL
18362986 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394785 Supplier Novus Biologicals Supplier No. NBP315715100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PLA2G2A Monoclonal antibody specifically detects PLA2G2A in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G2A
Applications Western Blot
Classification Monoclonal
Clone 3X9F5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 3.1.1.4, GIIC sPLA2, Group IIA phospholipase A2, MOM1, Non-pancreatic secretory phospholipase A2, NPS-PLA2, Phosphatidylcholine 2-acylhydrolase 2A, phospholipase A2, group IIA (platelets, synovial fluid), phospholipase A2, membrane associated, PLA2BPLA2, PLA2LPLA2S, PLAS1, RASF-A, sPLA2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 65-144 of human PLA2G2A (P14555).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5320
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.