missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G1B Antibody (Diaccurate patent anti-sPLA2-GIB) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28606JF669
This item is not returnable.
View return policy
Description
PLA2G1B Monoclonal antibody specifically detects PLA2G1B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PLA2G1B | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
| PLA2G1B | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5319 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Diaccurate patent anti-sPLA2-GIB | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction