missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G1B Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89682CL1
This item is not returnable.
View return policy
Description
PLA2G1B Monoclonal antibody specifically detects PLA2G1B in Human samples. It is validated for ELISA
Specifications
| PLA2G1B | |
| Monoclonal | |
| CoraFluor 1 | |
| Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PLA2G1B (Accession#: NP_000919.1; Met1-Ser148). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5319 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction