missing translation for 'onlineSavingsMsg'
Learn More

PLA2G1B Antibody (001), Alexa Fluor™ 594 , Novus Biologicals™

Product Code. 18415065 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415065 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415065 Supplier Novus Biologicals Supplier No. NBP289682AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PLA2G1B Monoclonal antibody specifically detects PLA2G1B in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G1B
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PLA2G1B (NP_000919.1; Met1-Ser148).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5319
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.