missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G16/HRASLS3 Antibody (OTI1A5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46429
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
PLA2G16/HRASLS3 Monoclonal specifically detects PLA2G16/HRASLS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PLA2G16/HRASLS3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:4000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| Adipose-specific phospholipase A2, AdPLAHRASLS3, EC 3.1.1.4, group XVI phospholipase A2, HRAS-like suppressor 3Ca-independent phospholipase A1/2, H-REV107-1, HREV107-3adipose-specific PLA2, HREV107H-rev 107 protein homolog, MGC118754, phospholipase A2, group XVI, Renal carcinoma antigen NY-REN-65 | |
| Mouse | |
| 17.8 kDa | |
| 0.1 mL | |
| Stem Cell Markers | |
| 11145 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1A5 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| PLA2G16 | |
| Full length human recombinant protein of human PLA2G16(NP_009000) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu