missing translation for 'onlineSavingsMsg'
Learn More

PLA2G16/HRASLS3 Antibody (4C4-2A11), Novus Biologicals™

Product Code. 18364479 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364479 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364479 Supplier Novus Biologicals Supplier No. H00011145M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLA2G16/HRASLS3 Monoclonal antibody specifically detects PLA2G16/HRASLS3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G16/HRASLS3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4C4-2A11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01387
Gene Alias Adipose-specific phospholipase A2, AdPLAHRASLS3, EC 3.1.1.4, group XVI phospholipase A2, HRAS-like suppressor 3Ca-independent phospholipase A1/2, H-REV107-1, HREV107-3adipose-specific PLA2, HREV107H-rev 107 protein homolog, MGC118754, phospholipase A2, group XVI, Renal carcinoma antigen NY-REN-65
Host Species Mouse
Immunogen PLA2G16 (AAH01387, 1 a.a. ~ 162 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRAPIPEPKPGDLIEIFRPFYRHWAIYVGDGYVVHLAPPSEVAGAGAASVMSALTDKAIVKKELLYDVAGSDKYQVNNKHDDKYSPLPCSKIIQRAEELVGQEVLYKLTSENCEHFVNELRYGVARSDQVRDVIIAASVAGMGLAAMSLIGVMFSRNKRQKQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11145
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.