missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G12A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16009-1-AP-150UL
This item is not returnable.
View return policy
Description
Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.Specifications
| PLA2G12A | |
| Polyclonal | |
| Unconjugated | |
| PLA2G12A | |
| GXII, GXII sPLA2, phospholipase A2, group XIIA, PLA2G12, PLA2G12A, ROSSY, sPLA2 XII, UNQ2519/PRO6012 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362039, 66350, 81579 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BZM1, Q9EPR2 | |
| Pla2g12a | |
| PLA2G12A Fusion Protein Ag8722 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction