missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2G12A Polyclonal Antibody
GREENER_CHOICE

Product Code. 13239458
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13239458 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13239458 Supplier Invitrogen™ Supplier No. PA522204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, H1299, HeLa, Raji. Predicted reactivity: Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G12A
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PLA2G12A
Gene Accession No. Q9BZM1
Gene Alias 2310004B05Rik; FKSG38; group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; group XIIA secretory phospholipase A2; GXII; GXII sPLA2; GXII-1; GXII-1-PLA2; mGXII-1; phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2 group XIIA; phospholipase A2, group XII; phospholipase A2, group XIIA; Pla2g12; PLA2G12A; Rossy; sPLA2-XII; UNQ2519/PRO6012
Gene Symbols PLA2G12A
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Phospholipase A2 XIIA. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.