missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA1A Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82024IR
This item is not returnable.
View return policy
Description
PLA1A Polyclonal antibody specifically detects PLA1A in Human, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PLA1A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 755 | |
| EC 3.1.1, EC 3.1.1.-, NMD, phosphatidylserine-specific phospholipase A1alpha, phospholipase A1 member A, PS-PLA1, PSPLA1Phosphatidylserine-specific phospholipase A1 | |
| PLA1A antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human PLA1A. The immunogen is located within amino acids 380 - 430 of PLA1A. Amino Acid Squence: KFKFQSSNRVWKKDRTT | |
| 0.1 mL | |
| Immunology | |
| 51365 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction