missing translation for 'onlineSavingsMsg'
Learn More

PLA1A Antibody - BSA Free, Novus Biologicals™

Product Code. 18342430 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342430 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342430 Supplier Novus Biologicals Supplier No. NBP282024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLA1A Polyclonal specifically detects PLA1A in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA1A
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias EC 3.1.1, EC 3.1.1.-, NMD, phosphatidylserine-specific phospholipase A1alpha, phospholipase A1 member A, PS-PLA1, PSPLA1Phosphatidylserine-specific phospholipase A1
Gene Symbols PLA1A
Host Species Rabbit
Immunogen PLA1A antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human PLA1A. The immunogen is located within amino acids 380 - 430 of PLA1A. Amino Acid Squence: KFKFQSSNRVWKKDRTT
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51365
Test Specificity At least three isoforms of PLA1A are known to exist; this antibody will detect all three isoforms.
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.