missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PKRA 7

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
PKRA 7 is a potent prokineticin (PK) receptor antagonist (IC50 values are 5.0 and 8.2 nM for PKR1 and PKR2, respectively). Reduces IL-1β and lL-6 expression in the joints and suppresses severity of arthritis in a mouse model of rheumatoid arthritis. Inhibits PK2-induced expression of pro-migratory chemokines and chemokine receptors in macrophages. Exhibits antitumor activity against glioblastoma and pancreatic cancer xenograft tumor models. Also exhibits antiangiogenic activity in vivo. Brain penetrant. Cell-permeable.
Specifications
Specifications
| Chemical Name or Material | (3 R)-N-[(9-Chloro-3,4-dihydro-2 H-1,5-benzodioxepin-7-yl)methyl]-1-[(4-fluoro-3-methoxyphenyl)methyl]-N-(2-methylpropyl)-3-pyrrolidinecarboxamide hydrochloride |
| Quantity | 5 mg |
| Target | Prokineticin R2/PROKR2 |
| Molecular Formula | C27H34ClFN2O4.HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction