missing translation for 'onlineSavingsMsg'
Learn More

PKR-IN-2, MedChemExpress

Product Code. 16487848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487848 10 mg 10mg
16487666 100 mg 100mg
16473797 50 mg 50mg
16420318 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487848 Supplier Medchem Express Supplier No. HY19702
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PKR-IN-2 is a pyruvate kinase isoform PKR activator extracted from patent WO2014139144A1, compound 160. PKR-IN-2 can be used for the research of PKR function related diseases, including cancer, diabetes, obesity, autoimmune disorders, and benign prostatic hyperplasia.

PKR-IN-2 is a pyruvate kinase isoform PKR activator extracted from patent WO2014139144A1, compound 160. PKR-IN-2 can be used for the research of PKR function related diseases, including cancer, diabetes, obesity, autoimmune disorders, and benign prostatic hyperplasia.

Specifications

Chemical Name or Material PKR-IN-2
CAS 1628428-01-2
Color White
Purity Grade Notes Research
Molecular Formula C24H28N4O4S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (106.71 mM; Need ultrasonic)
SMILES N1C2C(=C(S(NC3=CC=C(C(N4CCC(O)(CC(C)C)CC4)=O)C=C3)(=O)=O)C=CC=2)N=CC=1
Molecular Weight (g/mol) 468.57
Formula Weight 468.57
Percent Purity 95.14%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.