missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77266-0.025mg
This item is not returnable.
View return policy
Description
PKR Polyclonal specifically detects PKR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| PKR | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| Rabbit | |
| 68 kDa | |
| 0.025 mg | |
| Apoptosis, Signal Transduction | |
| 5610 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5-5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5-5 μg/mL | |
| AAP57628 | |
| EIF2AK2 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PKR. The immunogen is located within the last 50 amino acids of PKR. Amino Acid Squence: TVWKKSPEKNERHT | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human PKR has 2 isoforms, including isoform 1 (551aa, 62.1kD) and isoform S (510aa, 57.4kD. This antibody detects both human isoforms. Mouse PKR has 1 isoform (515aa, 58.3kD). Rat PKR has only one isoform identified so far (513aa, 58.3kD). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction