missing translation for 'onlineSavingsMsg'
Learn More

PKR Antibody (1D11), Novus Biologicals™

Product Code. 18339168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339168 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339168 Supplier Novus Biologicals Supplier No. H00005610M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKR Monoclonal antibody specifically detects PKR in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKR
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002750
Gene Alias double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated
Host Species Mouse
Immunogen EIF2AK2 (NP_002750, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAGDLSAGFFMEELNTYRQKQGVVLKYQELPNSGPPHDRRFTFQVIIDGREFPEGEGRSKKEAKNAAAKLAVEILNKEKKAVSPLLLTTTNSSEGLSMGN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5610
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.