Learn More
PKP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14062-1-AP-20UL
 View more versions of this product
Product Code. 16812534
  • £135.00 / 20µL
Description
Plays a role as a regulator of Rho activity during cytokinesis. May play a role in junctional plaques.
Specifications
Specifications
PKP4
Polyclonal
Unconjugated
PKP4
p0071, PKP4, plakophilin 4
Rabbit
Antigen Affinity Chromatography
RUO
227937, 295625, 8502
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FH0, Q99569
PKP4
PKP4 Fusion Protein Ag5113
20 μL
Primary
Rat, Mouse, Human