missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553144
This item is not returnable.
View return policy
Description
Immunogen sequence: VGNGNLHRTS SVPEYVYNLH LVENDFVGGR SPVPKTYDML KAGTTATYEG RWGRGTAQYS SQKSVEERSL RHPLRRLEIS PDSSPERAHY THSDYQYSQR SQAGHTLHHQ ESRRAALLVP PRY Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 41%.
Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
Specifications
| PKP2 | |
| Polyclonal | |
| Unconjugated | |
| PKP2 | |
| 1200008D14Rik; 1200012P04Rik; AA516617; ARVD9; PKP2; Pkp2l; plakophilin 2; plakophilin-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99959 | |
| PKP2 | |
| Recombinant protein corresponding to Human PKP2. Recombinant protein control fragment (Product #RP-89628). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction