missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKNOX1 Polyclonal Antibody

Description
PA1-9550 detects PKNOX1 in human samples. PA1-9550 has been successfully used in Western blot procedures. By Western blot, PA1-9550 detects a 65 kDa protein representing PKNOX1. PA1-9550 immunogen is purified recombinant PKNOX1.1.
PKNOX1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
Specifications
| Antigen | PKNOX1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Pknox1 |
| Gene Accession No. | P55347 |
| Gene Alias | D17Wsu76e; homeobox protein PKNOX1; Homeobox protein PREP-1; human homeobox-containing protein; Pbx regulating protein-1; Pbx/knotted 1 homeobox; PBX/knotted 1 homeobox 1; PBX/knotted homeobox 1; PKNOX1; pkonx1c; PREP1 |
| Gene Symbols | Pknox1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?