Learn More
Invitrogen™ Human PKN2 (PRK2), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length
Specifications
Specifications
| Accession Number | Q16513 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5586 |
| Molecular Weight (g/mol) | 141.5 kDa |
| Name | Human PKN2 (PRK2), GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | 6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; p140 kinase; PAK2; Pak-2; PKN gamma; PKN2; PRK2; PRKCL2; PRO2042; protease-activated kinase 2; protein kinase C-like 2; protein kinase N2; protein-kinase C-related kinase 2; RP5-905H16.1; serine/threonine kinase 7; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2-like; si:ch211-83f14.1; Stk7; wu:fa97c07 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.