missing translation for 'onlineSavingsMsg'
Learn More

PKMYT1 Antibody (2A3), Novus Biologicals™

Product Code. 18399358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399358 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399358 Supplier Novus Biologicals Supplier No. H00009088M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKMYT1 Monoclonal antibody specifically detects PKMYT1 in Human samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKMYT1
Applications Western Blot, ELISA, Proximity Ligation Assay, Sandwich ELISA
Classification Monoclonal
Clone 2A3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Proximity Ligation Assay, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004194
Gene Alias DKFZp547K1610, EC 2.7.11.1, FLJ20093, Myt1 kinase, MYT1membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase, protein kinase, membrane associated tyrosine/threonine 1
Host Species Mouse
Immunogen PKMYT1 (NP_004194.3, 400 a.a. ~ 499 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PASWLQPLGPPATPPGSPPCSLLLDSSLSSNWDDDSLGPSLSPEAVLARTVGSTSTPRSRCTPRDALDLSDINSEPPRGSFPSFEPRNLLSLFEDTLDPT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9088
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.