missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKM2, p Ser37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-13307
This item is not returnable.
View return policy
Description
PKM2 Polyclonal antibody specifically detects PKM2 in Human samples. It is validated for Western Blot
Specifications
| PKM2 | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK | |
| Carrier-protein conjugated synthetic peptide corresponding to residues around human PKM2 (phospho Ser37). The exact sequence is proprietary. | |
| 100 μg | |
| Cancer, Lipid and Metabolism | |
| 5315 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction