missing translation for 'onlineSavingsMsg'
Learn More

PKM2-IN-1, MedChemExpress

Product Code. 16493868
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493868 10 mg 10mg
16470976 200 mg 200mg
16446597 5 mg 5mg
16420247 50 mg 50mg
16404926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493868 Supplier Medchem Express Supplier No. HY103617
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PKM2-IN-1 is a pyruvate kinase M2 (PKM2) inhibitor with an IC50 of 2.95 μM.

PKM2-IN-1 is a pyruvate kinase M2 (PKM2) inhibitor with an IC50 of 2.95 μM.

Specifications

Chemical Name or Material PKM2-IN-1
CAS 94164-88-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H19NO2S2
Quantity 10 mg
Solubility Information DMSO : 320 mg/mL (926.25 mM; Need ultrasonic)
SMILES S=C(N1CCCCC1)SCC(C2=O)=C(C)C(C3=C2C=CC=C3)=O
Molecular Weight (g/mol) 345.48
Formula Weight 345.48
Percent Purity 99.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.