missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PKI (5-24)

Description
PKI (5-24) is a high affinity PKA inhibitor (Ki = 2.3 nM).
Specifications
Specifications
| Species | Human |
| Purity | 0.95 |
| CAS | 99534-03-9 |
| Content And Storage | Store at −20°C |
| Molecular Weight (g/mol) | 2222.4g/mol |
| Quantity | 1 mg |
| Sequence | TTYADFIASGRTGRRNAIHD |
| Specific Reactivity | High affinity PKA inhibitor (Ki = 2.3nM) |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?