missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PKHD1 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 5314 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for BC110-60480. For further blocking peptide related protocol, click here . |
| Gene Alias | polycystic kidney and hepatic disease 1 (autosomal recessive) |
| Gene Symbol | PKHD1 |
| Quantity | 0.1 mg |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?