missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKG drug G1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
PKG drug G1 targets C42 of PKG Iα. PKG drug G1 can couple to vasodilation and blood pressure lowering by a C42 PKG Iα-independent mechanism.
Specifications
Specifications
| Chemical Name or Material | PKG drug G1 |
| Physical Form | Solid |
| CAS | 374703-78-3 |
| Color | Reddish Brown |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C13H11N3OS |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 103 mg/mL (400.30 mM) |
| SMILES | O=C(/C(N1)=C/C2=C(C)NC3=C2C=CC=C3)NC1=S |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction