missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKC zeta Polyclonal Antibody
GREENER_CHOICE

Product Code. 11592480
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11592480 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11592480 Supplier Invitrogen™ Supplier No. PA517589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with endogenous levels of other PKC isoforms.

The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC zeta
Applications Western Blot
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PRKCZ
Gene Accession No. Q05513
Gene Alias 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; AI098070; aPKCzeta; atypical protein kinase C; C80388; kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; PKC2; Pkcz; PKC-ZETA; Prkcz; protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, zeta; r14-3-3; R74924; zetaPKC
Gene Symbols PRKCZ
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human PKC zeta.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5590
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.