missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC theta Antibody (453416), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4368K
This item is not returnable.
View return policy
Description
PKC theta Monoclonal antibody specifically detects PKC theta in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| PKC theta | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF | |
| 453416 | |
| Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EC 2.7.11, EC 2.7.11.13, MGC126514, MGC141919, nPKC-theta, PRKCT, protein kinase C theta type, protein kinase C, theta | |
| E. coli-derived recombinant human PKC theta , His580-Ser706, Accession # Q04759 | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 5588 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction