missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PKC Pan (Thr497) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15594041
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15594041 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594041 Supplier Invitrogen™ Supplier No. PA536757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous levels of PKC alpha, beta-I, beta-II, gamma, delta, epsilon, eta, zeta and theta isoforms only when phosphorylated at a residue homologous to threonine 497 of human PKC alpha. Purity is >95% by SDS-PAGE.

Members of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differ among tissues and PKC family members exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca2+.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKC Pan (Thr497)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Prkca
Gene Accession No. F1M7Y5, P05129, P05696, P05771, P09215, P09216, P09217, P16054, P17252, P20444, P23298, P24723, P28867, P41743, P63318, P63319, P68403, P68404, Q02111, Q02156, Q02956, Q04759, Q05513, Q05655, Q62074, Q64617, Q9WTQ0
Gene Alias 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; 2310021H13Rik; 5830406C15Rik; A130035A12Rik; A130082F03Rik; AAG6; aging-associated gene 6; AI098070; AI385711; AI427505; AI875142; ALPS3; aPKClambda; aPKC-lambda/iota; aPKCzeta; atypical protein kinase C; atypical protein kinase C-lambda/iota; AW494342; C80388; CVID9; D14Ertd420e; DXS1179E; MAY1; mKIAA4165; nPKC-delta; nPKC-epsilon; nPKC-eta; nPKC-iota; nPKC-theta; nPKC-zeta; PKC; PKC theta; PKC zeta; PKC[d]; PKC[e]; PKC-0; PKC2; Pkca; PKC-A; PKC-alpha; Pkcb; PKC-B; PKC-beta; Pkcc; PKCD; PKCdelta; Pkce; Pkcea; PKCepsilon; PKCG; PKCgamma; PKC-gamma; Pkch; PKCI; PKCL; PKC-L; PKClambda; Pkcq; PKCtheta; PKC-theta; Pkcz; PKC-ZETA; PRKACA; Prkc; PRKCA; Prkcb; Prkcb1; Prkcb2; Prkcc; Prkcd; Prkce; PRKCG; Prkch; PRKCI; PRKCL; PRKC-lambda/iota; Prkcq; PRKCT; Prkcz; protein kinase C alpha; protein kinase C alpha type; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C eta; protein kinase C eta type; protein kinase C gamma; protein kinase C gamma type; protein kinase C iota; protein kinase C iota type; protein kinase C theta; protein kinase C theta type; protein kinase C type I (gamma type); protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, alpha; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C, epsilon; protein kinase C, eta; protein kinase C, gamma; protein kinase C, iota; protein kinase C, lambda; protein kinase C, theta; protein kinase C, zeta; protein kinase C[d]; protein kinase c-alpha; protein kinase C-eta; r14-3-3; R74924; R75156; RATPKCI; SCA14; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; zetaPKC
Gene Symbols Prkca, PRKCB, PRKCD, PRKCE, PRKCG, Prkch, Prkci, PRKCQ, PRKCZ
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human PKC-pan around the phosphorylation site of Threonine 497.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170538, 18750, 18751, 18752, 18753, 18754, 18755, 18759, 18761, 18762, 24680, 24681, 25023, 25522, 29340, 5578, 5579, 5580, 5581, 5582, 5583, 5584, 5588, 5590, 81749, 84006, 85420
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.