missing translation for 'onlineSavingsMsg'
Learn More

PKC mu Rabbit anti-Human, Mouse, Rat, Clone: 2A5U1, Novus Biologicals™

Rabbit Monoclonal Antibody

£158.00 - £386.00

Specifications

Antigen PKC mu
Clone 2A5U1
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18324934
View Documents
Novus Biologicals
NBP3-16130-20UL
20 μg
£158.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18381064
View Documents
Novus Biologicals
NBP3-16130-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PKC mu Monoclonal antibody specifically detects PKC mu in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

PKC mu
Western Blot
Unconjugated
Rabbit
Golgi Apparatus Markers, Signal Transduction
PBS, 0.05% BSA, 50% glycerol, pH7.3
5587
IgG
Affinity purified
2A5U1
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 2.7.11, nPKC-D1, nPKC-mu, PKCM, PKC-MU, PKD1, PKDEC 2.7.11.13, PRKCMPKC-mu, Protein kinase C mu type, protein kinase C, mu, Protein kinase D, protein kinase D1, serine/threonine-protein kinase D1
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human PKC mu (Q15139).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.