missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC iota Antibody (PRKCI/4912) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14156JF525
This item is not returnable.
View return policy
Description
PKC iota Monoclonal antibody specifically detects PKC iota in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| PKC iota | |
| Monoclonal | |
| Janelia Fluor 525 | |
| aPKC-lambda/iota, Atypical protein kinase C-lambda/iota, DXS1179E, EC 2.7.11, EC 2.7.11.13, MGC26534, nPKC-iota, PKCI, PRKC-lambda/iota, protein kinase C iota type, protein kinase C, iota | |
| Recombinant fragment (around aa100-300) of human PKC iota protein (exact sequence is proprietary) (Uniprot: P41743) | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 5584 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| PRKCI/4912 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction