missing translation for 'onlineSavingsMsg'
Learn More

PKC iota Antibody (PRKCI/4911), HRP, Novus Biologicals™

Product Code. 18391547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391547 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391547 Supplier Novus Biologicals Supplier No. NBP314155H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKC iota Monoclonal antibody specifically detects PKC iota in Human, Canine, Feline samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC iota
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PRKCI/4911
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias aPKC-lambda/iota, Atypical protein kinase C-lambda/iota, DXS1179E, EC 2.7.11, EC 2.7.11.13, MGC26534, nPKC-iota, PKCI, PRKC-lambda/iota, protein kinase C iota type, protein kinase C, iota
Host Species Mouse
Immunogen Recombinant fragment (around aa100-300) of human PKC iota protein (exact sequence is proprietary) (Uniprot: P41743)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5584
Target Species Human, Canine, Feline
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.