missing translation for 'onlineSavingsMsg'
Learn More

PKC-IN-1, MedChemExpress

Product Code. p-200099631
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489727

Brand: Medchem Express HY16903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PKC-IN-1 is a potent, ATP-competitive and reversible inhibitor of conventional PKC enzymes with Kis of 5.3 and 10.4 nM for human PKCβ and PKCα, and IC50s of 2.3, 8.1, 7.6, 25.6, 57.5, 314, 808 nM for PKCα, PKCβI, PKCβII, PKCθ, PKCγ, PKC mu and PKCε, respectively.

PKC-IN-1 is a potent, ATP-competitive and reversible inhibitor of conventional PKC enzymes with Kis of 5.3 and 10.4 nM for human PKCβ and PKCα, and IC50s of 2.3, 8.1, 7.6, 25.6, 57.5, 314, 808 nM for PKCα, PKCβI, PKCβII, PKCθ, PKCγ, PKC mu and PKCε, respectively.

Specifications

Chemical Name or Material PKC-IN-1
Physical Form Solid
CAS 1046787-18-1
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H37FN8O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 25 mg/mL (49.94 mM)
SMILES O=C(N1[C@@H](C)CN(CC2CCOCC2)[C@H](C)C1)N(C3)C(C)(C)C4=C3C(NC5=NC(C)=NC=C5F)=NN4
Molecular Weight (g/mol) 500.61
Formula Weight 500.61
Percent Purity 98.79%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.