missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | PKC gamma |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603326
|
Novus Biologicals
NBP2-38728-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18184789
|
Novus Biologicals
NBP2-38728 |
0.1 mL |
£488.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKC gamma Polyclonal specifically detects PKC gamma in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PKC gamma | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P05129 | |
| 5582 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PPDRLVLASIDQADFQGFTYVNPDFVHPDARSPTSPVPVPVM | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.13, MGC57564, PKCCSCA14, PKC-gamma, PKCGprotein kinase C gamma type, protein kinase C, gamma | |
| PRKCG | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title