missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKC eta Recombinant Rabbit Monoclonal Antibody (24GB1480)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557192
This item is not returnable.
View return policy
Description
PKC eta Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Specifications
| PKC eta | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRKCH | |
| LOW QUALITY PROTEIN: protein kinase C eta type; MGC26269; MGC5363; nPKC-eta; PKC; Pkch; PKCL; PKC-L; PRKCH; PRKCL; protein kinase C eta; protein kinase C eta type; protein kinase C eta type pseudogene; protein kinase C theta; protein kinase C, eta; protein kinase C-eta; Unknown (protein for MGC:140784) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5583, 81749 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1480 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24723, Q64617 | |
| PRKCH | |
| A synthesized peptide derived from human PKC eta (aa 610-683). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction