missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC eta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-12447
This item is not returnable.
View return policy
Description
PKC eta Polyclonal antibody specifically detects PKC eta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| PKC eta | |
| Polyclonal | |
| Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.13, nPKC-eta, PKCLMGC26269, PKC-LMGC5363, PRKCLprotein kinase C eta type, protein kinase C, eta | |
| Synthetic peptide taken within amino acid region 1-50 on human PKC eta protein. Phosphorylated S28 & S32. | |
| 100 μg | |
| Protein Kinase, Signal Transduction | |
| 5583 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction