missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKC beta Monoclonal Antibody (K40051_4D2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556039
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: DVLIVLVRDA KNLVPMDPNG LSDPYVKLKL IPDPKSESKQ KTKTIKCSLN PEWNETFRFQ LKESDKDRRL SVEIWDWDLT SRNDFMGSLS FGISELQKAS VDGWFKLLSQ EEGEYFNVPV PPEGSEANEE LRQKFERAKI SQGTKVPEEK TTNTVSKFDN NGNRDRMKLT
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| PKC beta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P05771, P68403 | |
| PRKCB | |
| Recombinant human PRKCB (immunogen range is 171-340). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| K40051_4D2 | |
| Unconjugated | |
| PRKCB | |
| A130082F03Rik; PKCB; PKC-B; PKC-beta; PRKCB; PRKCB1; PRKCB2; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide | |
| Mouse | |
| Protein A | |
| RUO | |
| 25023, 5579 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction