missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC alpha, p Thr638 Antibody (S09-7C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19548-50ul
This item is not returnable.
View return policy
Description
PKC alpha Monoclonal antibody specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PKC alpha | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha | |
| A synthetic phosphopeptide corresponding to residues surrounding Thr638 of human PKC alpha (Uniprot # P17252) | |
| 50 μg | |
| Cancer, Cellular Markers, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 5578 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S09-7C7 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction