missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC alpha, p Ser226 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05437
This item is not returnable.
View return policy
Description
PKC alpha Polyclonal antibody specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PKC alpha | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha | |
| A synthetic phosphorylated peptide around S226 of human PKC alpha (NP_002728.2). NESFT | |
| 100 μg | |
| Cancer, Cellular Markers, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 5578 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction