missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC alpha Antibody (MC5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-201PE
This item is not returnable.
View return policy
Description
PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Fish, Rabbit, Sheep, Zebrafish samples. It is validated for Flow Cytometry.
Specifications
| PKC alpha | |
| Monoclonal | |
| PE | |
| P17252 | |
| PRKCA | |
| Purified bovine brain Protein Kinase C | |
| 0.1 mL | |
| Cancer, Cellular Markers, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 5578 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Fish, Rabbit, Sheep | |
| Purified |
| Flow Cytometry | |
| MC5 | |
| Flow Cytometry | |
| AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody is capable of detecting PKC beta as well. | |
| Store at 4C in the dark. | |
| IgG2a Kappa |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction