missing translation for 'onlineSavingsMsg'
Learn More

PKC alpha Antibody (MC5), DyLight 594, Novus Biologicals™

Product Code. 18418523 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418523 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418523 Supplier Novus Biologicals Supplier No. NB600201DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKC alpha Monoclonal antibody specifically detects PKC alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Fish, Baboon, Monkey, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Radioimmune Assays
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC alpha
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone MC5
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Host Species Mouse
Immunogen Purified bovine brain Protein Kinase C
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5578
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.