missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA R2 Antibody (OTI1F8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73428AF647
This item is not returnable.
View return policy
Description
PKA R2 Monoclonal specifically detects PKA R2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| PKA R2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRKAR2A | |
| Full length recombinant protein of human PKA R2 (NP_004148) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1F8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| protein kinase, cAMP-dependent, regulatory, type II, alpha, RII-alpha subunit | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific, Signal Transduction | |
| 5576.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction