missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX PKA Protein, Recombinant

Product Code. 17360174
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17360174

Brand: Eurofins DiscoverX 14440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant human full length PKA, catalytic subunit type alpha, untagged, for use in Kinase Assays.

Cyclic-AMP exerts its effects in animal cells mainly by activating the enzyme cyclic AMP-dependent protein kinase A (PKA). This enzyme catalyzes transfer of the terminal phosphate group from ATP to specific serines and threonines of selected proteins. Covalent phosphorylation of the appropriate amino acids of specific cellular proteins in turn results in altered cellullar function as a consequence of activation or inactivation of enzymes or altered membrane permeability to ions or other possible effects. PKA is found in all animal cells, and it is known that hormones or transmitters may be mediated by cAMP-stimulated phosphorylation of specific sites on neuronal or non-neuronal cells to exert their physiological function.

GenBank X07767

Specifications

Formulation mg/mL of enzyme in 30 mM potassium phosphate pH 7.4, 150 mM KCl, 1 mM EDTA, 1 mM DTT, 50% glycerol. Frozen solution.
Molecular Weight (g/mol) 41 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag Untagged
Expression System Recombinant enzyme expressed in E. coli cells
Protein Form Full-length
Purity or Quality Grade ≥59%
Protein PKA, catalytic subunit, recombinant
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.